2,4-Dinitro-5-fluorotoluene
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Fluoro-5-methyl-2,4-dinitrobenzene
ACTIAS25353G5EA
92
GBP
ACTIAS25353G5
2,4-Dinitro-5-fluorotoluene
2,4-Dinitro-5-fluorotoluene
Formula:
C₇H₅FN₂O₄ MW: 200.1240 g/mol |
MDL Number:
MFCD00205111 CAS Number: 349-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...