3-Amino-3-(4-methylphenyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: DL-3-Amino-3-p-tolyl-propionic acid3-Amino-3-p-tolyl-propionic acid

ACTIAS24739G5EA 32 GBP
ACTIAS24739G5 ACTIAS24739G25
3-Amino-3-(4-methylphenyl)propionic acid
3-Amino-3-(4-methylphenyl)propionic acid
Formula: C₁₀H₁₃NO₂
MW: 179.2157 g/mol
MDL Number: MFCD01863249
CAS Number: 68208-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR