Thiophene-2-ethylamine
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(2-Aminoethyl)thiophene
, 2-(Thiophen-2-yl)ethanamine
ACTIAS21271G25EA
0
GBP
ACTIAS21271G25
ACTIAS21271G100
Thiophene-2-ethylamine
Thiophene-2-ethylamine
Formula:
C₆H₉NS MW: 127.21 g/mol |
MDL Number:
MFCD00051495 CAS Number: 30433-91-1 EINECS: 250-196-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...