2,5-Thiophenedicarboxylic acid
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Thiophene-2,5-dicarboxylic acid
ACTIAS21752G25EA
13
GBP
ACTIAS21752G25
ACTIAS21752G100
2,5-Thiophenedicarboxylic acid
2,5-Thiophenedicarboxylic acid
Formula:
C₆H₄O₄S MW: 172.16 g/mol |
MDL Number:
MFCD00016896 CAS Number: 4282-31-9 EINECS: 224-284-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...