Methyl-4-ethynylbenzoate
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 4-ethynylbenzoate
ACTIAS23486G5EA
32
GBP
ACTIAS23486G5
Methyl-4-ethynylbenzoate
Methyl 4-ethynylbenzoate
Formula:
C₁₀H₈O₂ MW: 160.17 g/mol |
MDL Number:
MFCD00168820 CAS Number: 3034-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...