Methyl-4-methoxysalicylate
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-hydroxy-4-methoxybenzoate
ACTIAS22823G100EA
224
GBP
ACTIAS22823G25
ACTIAS22823G100
Methyl-4-methoxysalicylate
Methyl 4-methoxysalicylate
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol |
MDL Number:
MFCD00008424 CAS Number: 5446-02-6 EINECS: 226-657-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...