2-Phenoxybenzonitrile
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ACTIAS24083G1EA
64
GBP
ACTIAS24083G1
ACTIAS24083G5
2-Phenoxybenzonitrile
2-Phenoxybenzonitrile
Formula:
C₁₃H₉NO MW: 195.2167 g/mol |
MDL Number:
MFCD08271960 CAS Number: 6476-32-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...