3-Amino-3-(4-chlorophenyl)propionic acid

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Amino-3-(4-chlorophenyl)propanoic acid3-Amino-3-(p-chlorophenyl)propionic acid

ACTIAS22675G5EA 55 GBP
ACTIAS22675G5 ACTIAS22675G25
3-Amino-3-(4-chlorophenyl)propionic acid
3-Amino-3-(4-chlorophenyl)propionic acid
Formula: C₉H₁₀ClNO₂
MW: 199.64 g/mol
MDL Number: MFCD00126370
CAS Number: 19947-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR