4-Iodo-1H-pyrazole
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Iodopyrazole
ACTIAS21188G100EA
49
GBP
ACTIAS21188G100
ACTIAS21188G25
4-Iodo-1H-pyrazole
4-Iodo-1H-pyrazole
Formula:
C₃H₃IN₂ MW: 193.97 g/mol |
MDL Number:
MFCD00005244 CAS Number: 3469-69-0 EINECS: 222-434-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...