Methyl-3-aminopyrazine-2-carboxylate
Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-aminopyrazine-3-carboxylate
ACTIAS21729G25EA
0
GBP
ACTIAS21729G25
ACTIAS21729G5
ACTIAS21729G100
Methyl-3-aminopyrazine-2-carboxylate
Methyl 3-aminopyrazine-2-carboxylate
Formula:
C₆H₇N₃O₂ MW: 153.14 g/mol |
MDL Number:
MFCD00010097 CAS Number: 16298-03-6 EINECS: 240-387-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...