6-Bromo[1,2,4]triazolo[4,3-a]pyridine

Supplier: ACTIVATE SCIENTIFIC GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Bromo-[1,2,4]triazolo[4,3-a]pyridine

ACTIAS50584G5EA 150 GBP
ACTIAS50584G5 ACTIAS50584G1
6-Bromo[1,2,4]triazolo[4,3-a]pyridine
6-Bromo-[1,2,4]triazolo[4,3-a]pyridine
Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
MDL Number: MFCD08689506
CAS Number: 108281-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR