6-Hydroxy-5-nitronicotinic acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL669679-5GEA
37.3
GBP
SIAL669679-5G
SIAL669679-1G
6-Hydroxy-5-nitronicotinic acid, Sigma-Aldrich®
6-Hydroxy-5-nitronicotinic acid
Formula:
C₆H₄N₂O₅ MW: 184.1100 g/mol |
MDL Number:
MFCD02180840 CAS Number: 6635-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...