4-(1-Piperidinyl)aniline ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Piperidinoaniline
L06767.03EA
0
GBP
L06767.03
4-(1-Piperidinyl)aniline ≥97%
4-(1-Piperidinyl)aniline
Formula:
C₁₁H₁₆N₂ MW: 176.26 g/mol Boiling Pt: 140…142 °C (1 mmHg) Melting Pt: 26…28 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051688 CAS Number: 2359-60-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...