2-(Bromomethyl)-4-fluorophenylboronic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBBB-2706-1GEA
0
GBP
COBBBB-2706-1G
COBBBB-2706-5G
2-(Bromomethyl)-4-fluorophenylboronic acid
2-(Bromomethyl)-4-fluorophenylboronic acid
Formula:
C₇H₇BBrFO₂ |
MDL Number:
MFCD06659867 CAS Number: 850568-01-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...