5-Bromo-4-pentylpyrimidine

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBHC-6075-1GEA 0 GBP
COBBHC-6075-1G COBBHC-6075-5G
5-Bromo-4-pentylpyrimidine
5-Bromo-4-pentylpyrimidine
Formula: C₉H₁₃BrN₂
MDL Number: MFCD09800934
CAS Number: 951884-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR