5-Bromo-4-hexylpyrimidine

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBHC-6077-5GEA 0 GBP
COBBHC-6077-5G COBBHC-6077-1G
5-Bromo-4-hexylpyrimidine
5-Bromo-4-hexylpyrimidine
Formula: C₁₀H₁₅BrN₂
MDL Number: MFCD09800936
CAS Number: 951884-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR