5-Bromo-4-heptylpyrimidine
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6084-5GEA
94.97
GBP
COBBHC-6084-5G
COBBHC-6084-1G
5-Bromo-4-heptylpyrimidine
5-Bromo-4-heptylpyrimidine
Formula:
C₁₁H₁₇BrN₂ |
MDL Number:
MFCD09800939 CAS Number: 951884-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...