5-Bromo-4-isopropylpyrimidine
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBHC-6071-1GEA
0
GBP
COBBHC-6071-1G
COBBHC-6071-5G
5-Bromo-4-isopropylpyrimidine
5-Bromo-4-isopropylpyrimidine
Formula:
C₇H₉BrN₂ |
MDL Number:
MFCD09800930 CAS Number: 951884-28-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...