5-Bromo-4-isopentylpyrimidine
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5-Bromo-4-(3-methylbutyl)pyrimidine
COBBHC-6083-1GEA
0
GBP
COBBHC-6083-1G
COBBHC-6083-5G
5-Bromo-4-isopentylpyrimidine
5-Bromo-4-isopentylpyrimidine
Formula:
C₉H₁₃BrN₂ |
MDL Number:
MFCD09800938 CAS Number: 951884-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...