1,2-Diamino-9,10-anthraquinone

Supplier: Biotium
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


Synonyms: 1,2-Diaminoanthraquinone

00300.EA 170 GBP
00300.
1,2-Diamino-9,10-anthraquinone
1,2-Diamino-9,10-anthraquinone
Formula: C₁₄H₁₀N₂O₂
MDL Number: MFCD00001219
CAS Number: 1758-68-5
EINECS: 217-156-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR