4-Isopropylphenylhydrazine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL362395-1GEA
67.8
GBP
SIAL362395-1G
4-Isopropylphenylhydrazine hydrochloride, Sigma-Aldrich®
4-Isopropylphenylhydrazine hydrochloride
Formula:
C₉H₁₅ClN₂ MW: 186.69 g/mol |
MDL Number:
MFCD00013461 CAS Number: 118427-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...