5-Methylisophthalic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIAL641383-1GEA 37.2 GBP
SIAL641383-1G SIAL641383-5G
5-Methylisophthalic acid, Sigma-Aldrich®
5-Methylisophthalic acid
Formula: C₉H₈O₄
MW: 180.16 g/mol
MDL Number: MFCD00013986
CAS Number: 499-49-0
EINECS: 207-881-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR