1-Bromo-4-(trans-4-butylcyclohexyl)benzene
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBOS-7499-1GEA
23.74
GBP
COBBOS-7499-1G
COBBOS-7499-5G
COBBOS-7499-25G
1-Bromo-4-(trans-4-butylcyclohexyl)benzene
1-Bromo-4-(trans-4-butylcyclohexyl)benzene
Formula:
C₁₆H₂₃Br |
MDL Number:
MFCD06658187 CAS Number: 516510-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...