2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Hydroxy-2,3,5,6-tetrafluorobenzotrifluoride
M-Clarity™ quality level = MQ100
SIAL384070-1GEA
45.8
GBP
SIAL384070-1G
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol, Sigma-Aldrich®
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)phenol
Formula:
C₇HF₇O |
MDL Number:
MFCD00155984 CAS Number: 2787-79-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...