4-Chloro-2-fluoro-5-nitrobenzoic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4072-5GEA
0
GBP
COBBCA-4072-5G
COBBCA-4072-25G
COBBCA-4072-1G
4-Chloro-2-fluoro-5-nitrobenzoic acid
4-Chloro-2-fluoro-5-nitrobenzoic acid
Formula:
C₇H₃ClFNO MW: 219.55 g/mol |
MDL Number:
MFCD04115725 CAS Number: 35112-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...