2-Chloro-4-fluoro-5-nitrobenzoic acid

Supplier: COMBI-BLOCKS INC.
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

COBBCA-4018-1GEA 7.91 GBP
COBBCA-4018-1G COBBCA-4018-5G
2-Chloro-4-fluoro-5-nitrobenzoic acid
2-Chloro-4-fluoro-5-nitrobenzoic acid
Formula: C₇H₃ClFNO
MW: 219.55 g/mol
MDL Number: MFCD04115715
CAS Number: 114776-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR