Isobutyl-4-hydroxybenzoate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC14228900EA
225
GBP
EHERC14228900
Isobutyl-4-hydroxybenzoate
Isobutyl 4-hydroxybenzoate
Formula:
C₁₁H₁₄O₃ MW: 194,23 |
MDL Number:
MFCD00020167 CAS Number: 4247-02-3 EINECS: 224-208-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...