4,6-Dimethoxysalicylaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4,6-Dimethoxy-2-hydroxybenzaldehyde
, 2-Hydroxy-4,6-dimethoxybenzaldehyde
M-Clarity™ quality level = MQ100
SIAL138797-1GEA
53.2
GBP
SIAL138797-1G
4,6-Dimethoxysalicylaldehyde, Sigma-Aldrich®
4,6-Dimethoxysalicylaldehyde
Formula:
C₉H₁₀O₄ MW: 182.1733 g/mol |
MDL Number:
MFCD00003328 CAS Number: 708-76-9 EINECS: 211-904-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...