3-Ethoxysalicylaldehyde ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B20702.30EA
101.3
GBP
B20702.30
B20702.18
3-Ethoxysalicylaldehyde ≥97%
3-Ethoxysalicylaldehyde
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 263…264 °C Melting Pt: 66…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003323 CAS Number: 492-88-6 EINECS: 207-765-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...