4-Bromo-2-(2-methoxyethoxy)anisole
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBOT-1173-5GEA
0
GBP
COBBOT-1173-5G
4-Bromo-2-(2-methoxyethoxy)anisole
4-Bromo-2-(2-methoxyethoxy)anisole
Formula:
C₁₀H₁₃BrO₃ |
MDL Number:
MFCD14584686 CAS Number: 1132672-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...