1-Bromo-4-pentyloxybenzene ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A12614.06EA
45.2
GBP
A12614.06
1-Bromo-4-pentyloxybenzene ≥98%
1-Bromo-4-pentyloxybenzene
Formula:
C₁₁H₁₅BrO MW: 243.15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00156123 CAS Number: 30752-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...