Dicyclohexylphenylphosphine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Dicyclohexyl(phenyl)phosphine
M-Clarity™ quality level = MQ100
SIAL288284-1GEA
94.3
GBP
SIAL288284-1G
Dicyclohexylphenylphosphine, Sigma-Aldrich®
Dicyclohexylphenylphosphine
Formula:
C₁₈H₂₇P MW: 274.38 g/mol |
MDL Number:
MFCD00003854 CAS Number: 6476-37-5 EINECS: 229-334-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...