Dicyclohexylphenylphosphine 95%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
Dicyclohexyl(phenyl)phosphine
ACRO407080010EA
64.1
GBP
ACRO407080010
Dicyclohexylphenylphosphine 95%
Dicyclohexylphenylphosphine
Formula:
C₁₈H₂₇P MW: 274.38 g/mol Melting Pt: 54…57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003854 CAS Number: 6476-37-5 EINECS: 229-334-7 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 56 to 61 °C |
Titration with HClO4 | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...