3-Fluoro-4-iodobenzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

SIAL678899-1GEA 48.6 GBP
SIAL678899-1G
3-Fluoro-4-iodobenzonitrile, Sigma-Aldrich®
3-Fluoro-4-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
MDL Number: MFCD07782076
CAS Number: 887266-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR