3-Fluoro-4-iodobenzonitrile ≥97%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
H55141.03EA
30.9
GBP
H55141.03
H55141.MD
3-Fluoro-4-iodobenzonitrile ≥97%
3-Fluoro-4-iodobenzonitrile
Formula:
C₇H₃FIN MW: 247.01 g/mol Melting Pt: 113…117 °C Storage Temperature: Ambient |
MDL Number:
MFCD07782076 CAS Number: 887266-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...