Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyldiphenylphosphine ≥98%

Supplier: Alfa Aesar
Warning



Formula: C₁₄H₁₅P
MW: 214,24 g/mol
Boiling Pt: 130…132 °C (1 mmHg)
Density: 1,048
Storage Temperature: Ambient
MDL Number: MFCD00015170
CAS Number: 607-01-2
UN: 3278
ADR: 6.1,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 50 g A18292.18
Glass bottle for solids 10 g A18292.09
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula weight 214.25
CAS No. 607-01-2
Purity 98%
Odour/scent Stench
Boiling point 130-132°/1mm
Molecular formula CH3CH2P(C6H5)2
MDL No. MFCD00015170
UN No. 3278

WRITE A REVIEW

Need help?

  • Whatever you have to say, positive or negative, is important to us.
  • Providing feedback takes only seconds.
  • Your feedback will be made available to the public on the web.

VWR.com will publish your name as vwr.com user.
You can use this name, or edit and save a nickname.
We recommend authors use their first and last initial.

Please confirm the industry you work in

Example: Pharmaceutical

Please confirm your role

Example: Clinical Trial Assistant

Please confirm your location

Example: New York, NY

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty