3,6-Dichlorophthalic anhydride, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ200
SIAL359858-1GEA
278
GBP
SIAL359858-1G
3,6-Dichlorophthalic anhydride, Sigma-Aldrich®
3,6-Dichlorophthalic anhydride
Formula:
C₈H₂Cl₂O₃ MW: 217.01 g/mol |
MDL Number:
MFCD00042786 CAS Number: 4466-59-5 EINECS: 224-733-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...