2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H33505.03EA
60.5
GBP
H33505.03
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-5-methyl-4-(trifluoromethyl)benzoic acid
Formula:
C₉H₆F₄O₂ MW: 222.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19687178 CAS Number: 1323955-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...