3-Bromo-5-(trifluoromethyl)benzoic acid
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
COBBCA-4351-25GEA
27.7
GBP
COBBCA-4351-25G
COBBCA-4351-5G
COBBCA-4351-100G
3-Bromo-5-(trifluoromethyl)benzoic acid
3-Bromo-5-(trifluoromethyl)benzoic acid
Formula:
C₈H₄BrF₃O₂ MW: 269.02 g/mol |
MDL Number:
MFCD03412186 CAS Number: 328-67-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...