4'-Butoxyacetanilide, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL637688-1GEA
61.5
GBP
SIAL637688-1G
4'-Butoxyacetanilide, Sigma-Aldrich®
4'-Butoxyacetanilide
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD00027197 CAS Number: 23563-26-0 EINECS: 245-739-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...