3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid ≥97%
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H32721.MDEA
37.2
GBP
H32721.MD
H32721.03
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid ≥97%
3-(3-Methyl-5-(trifluoromethoxy)phenyl)propionic acid
Formula:
C₁₁H₁₁F₃O₃ MW: 248.20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09025376 CAS Number: 916420-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...