(R)-(+)-2-Benzyloxypropionic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(R)-2-(Benzyloxy)propanoic acid
L17205.22EA
398
GBP
L17205.22
L17205.14
L17205.06
(R)-(+)-2-Benzyloxypropionic acid ≥98%
(R)-(+)-2-Benzyloxypropionic acid
Formula:
C₁₀H₁₂O₃ MW: 180.20 g/mol Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD06799065 CAS Number: 100836-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...