Pentachlorothioanisole
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC15120000EA
189
GBP
EHERC15120000
Pentachlorothioanisole
Pentachlorothioanisole
Formula:
C₇H₃Cl₅S |
MDL Number:
MFCD00013622 CAS Number: 1825-19-0 EINECS: 217-363-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...