2-Bromo-4-iodotoluene
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Bromo-4-iodo-1-methylbenzene
COBBHC-2382-5GEA
0
GBP
COBBHC-2382-5G
2-Bromo-4-iodotoluene
2-Bromo-4-iodotoluene
Formula:
C₇H₆BrI MW: 296.9300 g/mol |
MDL Number:
MFCD07779014 CAS Number: 26670-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...