Diisopropyl phthalate
Supplier: EHRENSTORFER GMBH
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EHERC16173700EA
144
GBP
EHERC16173700
Diisopropyl phthalate
Diisopropyl phthalate
Formula:
C₁₄H₁₈O₄ |
MDL Number:
MFCD00053717 CAS Number: 605-45-8 EINECS: 210-086-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...