(S)-α-(2-Oxiranyl)anisole, Sigma-Aldrich®
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(S)-Glycidyl phenyl ether
M-Clarity™ quality level = MQ100
SIAL04855-5G-FEA
172
GBP
SIAL04855-5G-F
(S)-α-(2-Oxiranyl)anisole, Sigma-Aldrich®
(S)-α-(2-Oxiranyl)anisole
Formula:
C₉H₁₀O₂ MW: 150.1745 g/mol |
MDL Number:
MFCD06659026 CAS Number: 71031-03-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...