4-Thioanisolemagnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
SIAL561762-50MLEA
295
GBP
SIAL561762-50ML
4-Thioanisolemagnesium bromide 0.5 mol/l in THF, Sigma-Aldrich®
4-Thioanisolemagnesium bromide
Formula:
C₇H₇BrMgS |
CAS Number:
18620-04-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...