2-(Hydroxymethyl)-9,10-anthraquinone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL226521-1GEA
48
GBP
SIAL226521-1G
2-(Hydroxymethyl)-9,10-anthraquinone, Sigma-Aldrich®
2-(Hydroxymethyl)-9,10-anthraquinone
Formula:
C₁₅H₁₀O₃ |
MDL Number:
MFCD00001236 CAS Number: 17241-59-7 EINECS: 241-274-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...