1-Amino-4-bromo-2-methyl-9,10-anthraquinone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIAL246697-5GEA
148
GBP
SIAL246697-5G
1-Amino-4-bromo-2-methyl-9,10-anthraquinone, Sigma-Aldrich®
1-Amino-4-bromo-2-methyl-9,10-anthraquinone
Formula:
C₁₅H₁₀BrNO₂ |
MDL Number:
MFCD00001221 CAS Number: 81-50-5 EINECS: 201-355-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...