2,2'-Bithiophene-5-boronic acid pinacol ester
Supplier: COMBI-BLOCKS INC.
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-([2,2'-Bithiophen]-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
COBBPN-8634-1GEA
27.7
GBP
COBBPN-8634-1G
2,2'-Bithiophene-5-boronic acid pinacol ester
2,2'-Bithiophene-5-boronic acid pinacol ester
Formula:
C₁₄H₁₇BO₂S₂ MW: 292.2 g/mol |
MDL Number:
MFCD04039974 CAS Number: 479719-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...